Learn More
Ibidi USA Luer Lock Connector Female
SDP
Catalog No. NC0844130
Encompass
Luer Lock Connector Female connects male Luers with 0.5, 0.8, or 1.6mm ID tubing. polypropylene, sterile, 25 pcs./box
This item is not returnable. View return policy
Catalog No. NC0844130 Supplier Ibidi USA Supplier No. 10825
Luer Lock Connector Female connects male Luers with 0.5, 0.8, or 1.6mm ID tubing. polypropylene, sterile, 25 pcs./box