Learn More
Anton Paar Female Luer Adapter 1/4"""""""" UNF
Catalog No. NC0164234
Encompass_Preferred
Converts female 1/4"""" UNF thread into female luer for use with syringesSuitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230
Catalog No. NC0164234 Supplier Anton Paar Supplier No. 64792
Converts female 1/4"""" UNF thread into female luer for use with syringesSuitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230