Learn More
Chemglass Life Sciences Female Luer Adapter, 1/4"-28 Male Thread
SDP
Catalog No. CG103715
Encompass_Preferred
Female Luer Adapter, 1/4"-28 Male Thread. Replacement female luer adapter for item CG103710. Complete adapter (CG103710) is supplied with 90¿ valve, 3-port 24/40 PTFE Adapter and 10ft of 1/8¿ OD PTFE tubing.
Catalog No. CG103715 Supplier Chemglass Life Sciences Supplier No. CG103715
Female Luer Adapter, 1/4"-28 Male Thread. Replacement female luer adapter for item CG103710. Complete adapter (CG103710) is supplied with 90¿ valve, 3-port 24/40 PTFE Adapter and 10ft of 1/8¿ OD PTFE tubing.