Learn More
Trajan Scientific Female Luer Adapter, Female Luer to 1/16" Fitting, Provides Direct Syringe Connection to ZDV Fittings and Valves
Catalog No. 501090227
Encompass
Female Luer Adapter, Female Luer to 1/16" Fitting, Provides Direct Syringe Connection to ZDV Fittings and Valves
Catalog No. 50-109-0227 Supplier Trajan Scientific Supplier No. ZLA1
Female Luer Adapter, Female Luer to 1/16" Fitting, Provides Direct Syringe Connection to ZDV Fittings and Valves