Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Female Luer x Cap; 25/PK
Catalog No. NC0066724
Encompass
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.
Catalog No. NC0066724 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550228
Easily complete your luer fittings systems with these premium connectors. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available.