Learn More
Cole-Parmer Luer Fitting, Straight Union, CrystalVu™, Cleanroom Packed, Female Luer; 10/Pk
Catalog No. 502558074 Shop All Cole-Parmer Products
Encompass_Preferred
These Cole-Parmer cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. This straight union features a female luer coupler on each end to connect two male luer fittings together. Unions are available in five different formulations—select one that best suits your application.
Catalog No. 50-255-8074 Supplier Cole-Parmer Supplier No. 5011421
These Cole-Parmer cleanroom fittings are high-quality PVC-free fittings, ideal in biomedical, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. This straight union features a female luer coupler on each end to connect two male luer fittings together. Unions are available in five different formulations—select one that best suits your application.