Learn More
Chemglass Life Sciences Female Luer Fitting with 1/8" Barb, Polycarbonate
SDP
Catalog No. 5000083195
Encompass
The Female Luer thread style to 1/8 barb (3.2mm) ID Tubing is gamma stable and polycarbonate.Ideal for use with CLS-1399-F50.
Catalog No. 50-000-83195 Supplier Chemglass Life Sciences Supplier No. CLS1399G50
The Female Luer thread style to 1/8 barb (3.2mm) ID Tubing is gamma stable and polycarbonate.Ideal for use with CLS-1399-F50.