Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Female Luer to Hose Barb Adapter, 1/8"" ID; 25/PK
Catalog No. NC0380952
Encompass
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
Catalog No. NC0380952 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080008
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.