Learn More
Ibidi USA Female Luer Lock Coupler
SDP
Catalog No. NC1070285
Encompass
Female Luer Lock Coupler for connecting two male Luer Lock adapters. polypropylene, sterile, 25 pcs./box
This item is not returnable. View return policy
Catalog No. NC1070285 Supplier Ibidi USA Supplier No. 10823
Female Luer Lock Coupler for connecting two male Luer Lock adapters. polypropylene, sterile, 25 pcs./box