Learn More
AVANTOR MASTERFLEX (VWR LLC) Saint Gobain T-LL075 Sanitary Tubing Adapter; Polypropylene; 3/4"" Mini to Female Luer Lock
Catalog No. NC9664394
Encompass
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.
Catalog No. NC9664394 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3120050
Standard connection type for syringes, plus ideal for many applications with small tubing. These Female luer lock x 3/4" mini sanitary adapters are available in either polypropylene or PVDF.