Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence 316 SS Fitting; Female Luer x 1/4 to 5/16"" Hose Barb; 1/Ea
Catalog No. NC1488082
Encompass
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.
Catalog No. NC1488082 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150730
Exceptional durability. Easy to disassemble for cleaning. Ideal for harsh chemical environments. Autoclavable.