Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Straight, Female Luer to Barb Hose Adapter, 3/16"" ID; 25/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4550108
Encompass
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Catalog No. NC0157482