Learn More
Bellco Glass, Inc. Female Quick Turn Coupling For 1/4 Tubing
SDP
Catalog No. 501946381
Encompass_Preferred
Female Quick Turn Coupling ¼". Plastic quick turn tube couplingMaterial: clear polycarbonateMaterial meets: ANSI/HIMA md 70.1 & iso 594/1-1986Max pressure: 125psi @72°fTemperature range: 200°f (continuous)Use with air & waterTubing: use PVC, polyurethane, & silicone tubing with a durometer-of a75-a85Clamps: not requiredTechnical:Also known as lure lock fittings. It only takes a quarter-turn to connect and disconnect these couplings. Suitable for radiation sterilization. Not rated for vacuum applications. Can be connect to the male quick turn (7808-00003). Polycarbonate: fair chemical, impact & abrasion resistance. Fair strength.. MADE IN THE USA
Catalog No. 50-194-6381 Supplier Bellco Glass, Inc. Supplier No. 780800002
Female Quick Turn Coupling ¼". Plastic quick turn tube couplingMaterial: clear polycarbonateMaterial meets: ANSI/HIMA md 70.1 & iso 594/1-1986Max pressure: 125psi @72°fTemperature range: 200°f (continuous)Use with air & waterTubing: use PVC, polyurethane, & silicone tubing with a durometer-of a75-a85Clamps: not requiredTechnical:Also known as lure lock fittings. It only takes a quarter-turn to connect and disconnect these couplings. Suitable for radiation sterilization. Not rated for vacuum applications. Can be connect to the male quick turn (7808-00003). Polycarbonate: fair chemical, impact & abrasion resistance. Fair strength.. MADE IN THE USA