Learn More
Checkers Female Ramp for 5-Channel Yellow Jacket™ AMS Modular Accessibility System
Shop All Justrite Safety Group ProductsDescription
- Protects cables and hoses up to 1.25 in. outside diameter
- Connect unlimited center sections and add ramps
- Hinged lid for easy cable placement
- Patented technology with modular interlocking design
Specifications
Specifications
| Color | Yellow |
| Product Type | Cable Protector Kit |
| Material | HDPE |