Learn More
AVANTOR MASTERFLEX (VWR LLC) Cadence Luer Fitting, nickel-plated brass, female luer lock to female luer lock; 1/ea
Catalog No. NC0816015
Encompass
Exceptional durability. Easy to disassemble for cleaning. Autoclavable.
Catalog No. NC0816015 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4150756
Exceptional durability. Easy to disassemble for cleaning. Autoclavable.