Learn More
SAI Infusion Technologies Rat Femoral Artery Catheter - 71.8cm, for Rats 125g+
SDP
Catalog No. NC0039934
Encompass
Femoral Artery Catheter for Rats 125g or larger. 71.8cm catheter with a 7.8cm 2Fr PU tip, PE body and 4 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle. Product is for laboratory animal research only.
Catalog No. NC0039934 Supplier SAI Infusion Technologies Supplier No. RFACPE50X25
Femoral Artery Catheter for Rats 125g or larger. 71.8cm catheter with a 7.8cm 2Fr PU tip, PE body and 4 suture beads to anchor the catheter in place. Sterile. Access with a 22G blunt needle. Product is for laboratory animal research only.