Learn More
SAI Infusion Technologies Rat Femoral Artery Catheter - for Rats 150g +
SDP
Catalog No. NC0534569
Encompass
Femoral Vein Catheter for Rats 150g or larger. 79cm Tapered PU with a 4cm 2.3 Fr tip, and 3 suture beads to anchor the catheter in place. Sterile. Tapered design permits entry into the smaller, muscular arteries while maintaining strength and durability at the externalization point. Access with 22G blunt needle. For rat > 150g. Product is for laboratory animal research only.
Catalog No. NC0534569 Supplier SAI Infusion Technologies Supplier No. RFA01
Femoral Vein Catheter for Rats 150g or larger. 79cm Tapered PU with a 4cm 2.3 Fr tip, and 3 suture beads to anchor the catheter in place. Sterile. Tapered design permits entry into the smaller, muscular arteries while maintaining strength and durability at the externalization point. Access with 22G blunt needle. For rat > 150g. Product is for laboratory animal research only.