Learn More
Med Vet International Femoral Disarticulator, Large, 7.75", J0233B
Catalog No. 5000585395
Encompass_Preferred
Designed to disarticulate the head of the femur from the acetabulum. The curved blade slides around and between the head of the femur and acetabulum. The sharpened notch at the tip locates and cuts through the round ligament.
Catalog No. 50-005-85395 Supplier Med Vet International Supplier No. JOR233B
Designed to disarticulate the head of the femur from the acetabulum. The curved blade slides around and between the head of the femur and acetabulum. The sharpened notch at the tip locates and cuts through the round ligament.