Learn More
SAI Infusion Technologies Rat Femoral Vein Catheter - 71.7cm PE and PU, for Rats 125g+
SDP
Catalog No. NC0039936
Encompass
Femoral Vein Catheter for Rats 125g or larger. 71.7cm PE catheter with a 7.7cm PU tip and 4 suture beads to anchor the catheter in place. Sterile. Access witn a 22G blunt needle. Product is for laboratory animal research only.
Catalog No. NC0039936 Supplier SAI Infusion Technologies Supplier No. RFVCPE50X25
Femoral Vein Catheter for Rats 125g or larger. 71.7cm PE catheter with a 7.7cm PU tip and 4 suture beads to anchor the catheter in place. Sterile. Access witn a 22G blunt needle. Product is for laboratory animal research only.