Learn More
Zymo Research Corporation Femto™ Bacterial DNA Quantification Kit
SDP
Catalog No. NC0801485
Encompass_Preferred
Bacterial DNA can be reliably quantified in a background of non-bacterial DNA such as fungal, animal, and plant DNA, etc. This is essential for downstream applications that require accurate DNA input including quantifying bacteria DNA template for Next-generation sequencing library preparation and metagenomic analysis amongst others. With the Femto™ BacterialDNA Quantification Kit, one can dependably quantify as little as 20 fg DNA from 1 l purified from biological liquids, bacterial cultures, or environmental DNA samples.
Catalog No. NC0801485 Supplier Zymo Research Corporation Supplier No. E2006
Bacterial DNA can be reliably quantified in a background of non-bacterial DNA such as fungal, animal, and plant DNA, etc. This is essential for downstream applications that require accurate DNA input including quantifying bacteria DNA template for Next-generation sequencing library preparation and metagenomic analysis amongst others. With the Femto™ BacterialDNA Quantification Kit, one can dependably quantify as little as 20 fg DNA from 1 l purified from biological liquids, bacterial cultures, or environmental DNA samples.