Learn More
Zymo Research Corporation Femto™ Fungal DNA Quantification Kit
SDP
Catalog No. NC1150242
Encompass_Preferred
Fungal DNA can be reliably quantified in a background of non-fungal DNA such as bacterial, animal, and plant DNA, etc. This isessential for downstream applications that require accurate fungal DNA input including Next-Gen sequencing and metagenomic analysis. With the Femto™ Fungal DNA Quantification Kit, one can dependably quantify as little as 20 fg of fungal DNA in 1 l purified biological liquids, fungal cultures, or environmental DNA samples.
Catalog No. NC1150242 Supplier Zymo Research Corporation Supplier No. E2007
Fungal DNA can be reliably quantified in a background of non-fungal DNA such as bacterial, animal, and plant DNA, etc. This isessential for downstream applications that require accurate fungal DNA input including Next-Gen sequencing and metagenomic analysis. With the Femto™ Fungal DNA Quantification Kit, one can dependably quantify as little as 20 fg of fungal DNA in 1 l purified biological liquids, fungal cultures, or environmental DNA samples.