Learn More
Zymo Research Corporation Femto™ Human DNA Quantification Kit
SDP
Catalog No. NC0852033
Encompass_Preferred
Human DNA can be reliably quantified in a background of non-human DNA such as bacterial, fungal, animal, and plant DNA,etc. This is essential for downstream applications that require accurate DNA input including STR analysis amongst others. With the Femto™ Human DNA Quantification Kit, one can dependably quantify as little as 20 fg DNA from 1 l purified from biologicalliquids, anthropological, or forensic DNA samples.
Catalog No. NC0852033 Supplier Zymo Research Corporation Supplier No. E2005
Human DNA can be reliably quantified in a background of non-human DNA such as bacterial, fungal, animal, and plant DNA,etc. This is essential for downstream applications that require accurate DNA input including STR analysis amongst others. With the Femto™ Human DNA Quantification Kit, one can dependably quantify as little as 20 fg DNA from 1 l purified from biologicalliquids, anthropological, or forensic DNA samples.