Learn More
Aviva Systems Biology Corp FEN1 Antibody - C-terminal region (ARP46097_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46097_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FEN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FEN1. Alternate names: FEN-1, MF1, RAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59224
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.