Learn More
Aviva Systems Biology Corp FEN1 Antibody - N-terminal region (ARP46096_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46096_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FEN1. It was validated on Western Blot. . Alternate names: FEN-1, MF1, RAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-239