Learn More
TARGETMOL CHEMICALS INC FEN1-IN-3 5MG
SDP
Catalog No. 502579017
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FEN1-IN-3 is a human flap endonuclease-1 ( hFEN1 ) inhibitor that stabilizes hFEN1 with an EC 50 of 6.8 uM . purity: 98%
Catalog No. 50-257-9017 Supplier TARGETMOL CHEMICALS INC Supplier No. T91475MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FEN1-IN-3 is a human flap endonuclease-1 ( hFEN1 ) inhibitor that stabilizes hFEN1 with an EC 50 of 6.8 uM . purity: 98%