Learn More
Selleck Chemical LLC FEN1-IN-4 S3397-25mg
SDP
Catalog No. 503109314
Encompass_Preferred
FEN1-IN-4 (FEN1 Inhibitor C2 JUN93587 Compound 2) is an inhibitor of human flap endonuclease-1 (hFEN1) with IC50 of 30 nM for hFEN1-336
Catalog No. 50-310-9314 Supplier Selleck Chemical LLC Supplier No. S339725MG
Add to Cart
Add to Cart
FEN1-IN-4 (FEN1 Inhibitor C2 JUN93587 Compound 2) is an inhibitor of human flap endonuclease-1 (hFEN1) with IC50 of 30 nM for hFEN1-336