Learn More
Medchemexpress LLC Fen1-IN-SC13 | 2098776-03-3 | 98.4% | C24H23N3O3S | 100 MG
SDP
Catalog No. 5000354247
Encompass_Preferred
FEN1-IN-SC13 is a potent DNA fragmentation endonuclease 1 (FEN1) inhibitor that exhibits antitumor activity. It interferes with DNA replication and repair both in vitro and in cells. This compound has a high purity and specific molecular properties.
  • Potent FEN1 inhibitor with antitumor activity
  • Interferes with DNA replication and repair
  • High purity of 98.42%
  • Molecular weight of 433.52
  • Solid appearance, off-white to light yellow color
Catalog No. 50-003-54247 Supplier Medchemexpress LLC Supplier No. HY145758100MG
FEN1-IN-SC13 is a potent DNA fragmentation endonuclease 1 (FEN1) inhibitor that exhibits antitumor activity. It interferes with DNA replication and repair both in vitro and in cells. This compound has a high purity and specific molecular properties.
  • Potent FEN1 inhibitor with antitumor activity
  • Interferes with DNA replication and repair
  • High purity of 98.42%
  • Molecular weight of 433.52
  • Solid appearance, off-white to light yellow color