Learn More
Crescent Chemical Co Inc FENAMIPHOS SOLN 1 ML
SDP
Catalog No. 5080356
Encompass_Preferred
Fenamiphos 100 µg/mL in Acetonitrile Insecticide (WS solution) CAS 22224-92-6 Molecular Weight - 303.3574 Storage - RT Catalog Num - DRE-XA13420000AL pesticide
Catalog No. 50-803-56 Supplier Crescent Chemical Co Inc Supplier No. PSXA13420000AL
Fenamiphos 100 µg/mL in Acetonitrile Insecticide (WS solution) CAS 22224-92-6 Molecular Weight - 303.3574 Storage - RT Catalog Num - DRE-XA13420000AL pesticide