Learn More
Crescent Chemical Co Inc FENAMIPHOS-SULFOXIDE 100 MG
SDP
Catalog No. 5073857
Encompass_Preferred
Fenamiphos-sulfoxide Insecticide (Metabolite neat) CAS 31972-43-7 Molecular Weight - 319.3568 Storage - -18°C Catalog Num - DRE-C13422000 chemical
Catalog No. 50-738-57 Supplier Crescent Chemical Co Inc Supplier No. PSC13422000
Fenamiphos-sulfoxide Insecticide (Metabolite neat) CAS 31972-43-7 Molecular Weight - 319.3568 Storage - -18°C Catalog Num - DRE-C13422000 chemical