Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Fenbendazole

Catalog No. AC462471000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462471000 100 g
AC462470250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462471000 Supplier Thermo Scientific Chemicals Supplier No. 462471000

missing translation for 'validMainframeMsg'

CAS: 43210-67-9 | C15H13N3O2S | 299.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43210-67-9
Molecular Formula C15H13N3O2S
Molecular Weight (g/mol) 299.35
InChI Key HDDSHPAODJUKPD-UHFFFAOYSA-N
IUPAC Name methyl N-[6-(phenylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1

Specifications

Quantity 100 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in dmso
Formula Weight 299.35
Chemical Name or Material Fenbendazole
EINECSNumber 256-145-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.