Learn More
Cayman Chemical Fenbendazole, Purity: ≥98%, CAS: 43210-67-9, 10 g
SDP
Catalog No. 502984973
Encompass_Preferred
A benzimidazole anthelmintic; active against Giardia in vitro (IC50 = 0.3 μM); prevents infiltration of parasites into the brain in a rabbit model of E. cuniculi infection at 20 mg/kg; activates HIF-1α and prevents oxidative stress-induced death in primary neurons in vitro
Catalog No. 50-298-4973 Supplier Cayman Chemical Supplier No. 1968710 G
A benzimidazole anthelmintic; active against Giardia in vitro (IC50 = 0.3 μM); prevents infiltration of parasites into the brain in a rabbit model of E. cuniculi infection at 20 mg/kg; activates HIF-1α and prevents oxidative stress-induced death in primary neurons in vitro