Learn More
Medchemexpress LLC Fenbendazole-d3 | 1228182-47-5 | 99.7% | 1 MG
SDP
Catalog No. 5000409060
Encompass_Preferred
Fenbendazole-d3 is a deuterium-labeled Fenbendazole, functioning as a HIF-1α agonist that activates the HIF-1α-related GLUT1 pathway. Fenbendazole is an orally active benzimidazole anthelmintic agent with a broad antiparasitic range. It acts as a microtubule destabilizing agent, inducing cell-cycle arrest and mitotic cell death, and has demonstrated antitumor activity in mice xenografted with wild-type p53.
  • Deuterium labeled Fenbendazole.
  • HIF-1α agonist, activating the HIF-1α-related GLUT1 pathway.
  • Orally active benzimidazole anthelmintic agent with a broad antiparasitic range.
  • Microtubule destabilizing agent.
  • Causes cell-cycle arrest and mitotic cell death.
  • Exhibits antitumor activity in mice xenografted with wild-type p53.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09060 Supplier Medchemexpress LLC Supplier No. HYB0413S1MG
Fenbendazole-d3 is a deuterium-labeled Fenbendazole, functioning as a HIF-1α agonist that activates the HIF-1α-related GLUT1 pathway. Fenbendazole is an orally active benzimidazole anthelmintic agent with a broad antiparasitic range. It acts as a microtubule destabilizing agent, inducing cell-cycle arrest and mitotic cell death, and has demonstrated antitumor activity in mice xenografted with wild-type p53.
  • Deuterium labeled Fenbendazole.
  • HIF-1α agonist, activating the HIF-1α-related GLUT1 pathway.
  • Orally active benzimidazole anthelmintic agent with a broad antiparasitic range.
  • Microtubule destabilizing agent.
  • Causes cell-cycle arrest and mitotic cell death.
  • Exhibits antitumor activity in mice xenografted with wild-type p53.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.