Learn More
Medchemexpress LLC Fenbendazole-d3 | 1228182-47-5 | MFCD16652570 | 99.7% | 302.37 g/mol | C15H10D3N3O2S | 10 MG
SDP
Catalog No. 5000298948
Encompass_Preferred
Fenbendazole-d3 is a deuterium-labeled analogue of fenbendazole used as an analytical reference standard and tracer in research. It is applied in pharmacokinetic and metabolism studies and shows microtubule destabilizing and antiparasitic activity, with reported antitumor effects in preclinical models.
  • Deuterium-labeled reference standard for analytical studies.
  • High purity: 99.7% as supplied.
  • Suitable for pharmacokinetic and metabolism tracing.
  • Exhibits microtubule destabilizing activity and antiparasitic effects.
  • Available in small quantities for laboratory use (e.g., 10 mg).
Catalog No. 50-002-98948 Supplier Medchemexpress LLC Supplier No. HYB0413S10MG
Fenbendazole-d3 is a deuterium-labeled analogue of fenbendazole used as an analytical reference standard and tracer in research. It is applied in pharmacokinetic and metabolism studies and shows microtubule destabilizing and antiparasitic activity, with reported antitumor effects in preclinical models.
  • Deuterium-labeled reference standard for analytical studies.
  • High purity: 99.7% as supplied.
  • Suitable for pharmacokinetic and metabolism tracing.
  • Exhibits microtubule destabilizing activity and antiparasitic effects.
  • Available in small quantities for laboratory use (e.g., 10 mg).