Learn More
Apexbio Technology LLC FENBENDAZOLE-D3-1MG
SDP
Catalog No. 5000570383
Encompass_Preferred
Fenbendazole-d3 (CAS No 1228182-47-5) is a deuterium-labeled variant of fenbendazole used primarily in biomedical research particularly in the study of parasitic infections and cancer Originating from the benzimidazole class of compounds its mechanism of action involves the disruption of microtubule formation by binding to tubulin leading to the inhibition of cell division In vitro studies often demonstrate its efficacy in the nanomolar to low micromolar range It is commonly applied in various cell models and animal studies serving as a tool in drug screening processes to evaluate antiparasitic or antitumor activities Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70383 Supplier Apexbio Technology LLC Supplier No. SL10571
Fenbendazole-d3 (CAS No 1228182-47-5) is a deuterium-labeled variant of fenbendazole used primarily in biomedical research particularly in the study of parasitic infections and cancer Originating from the benzimidazole class of compounds its mechanism of action involves the disruption of microtubule formation by binding to tubulin leading to the inhibition of cell division In vitro studies often demonstrate its efficacy in the nanomolar to low micromolar range It is commonly applied in various cell models and animal studies serving as a tool in drug screening processes to evaluate antiparasitic or antitumor activities Experimental concentrations typically depend on the specific research design