Learn More
Medchemexpress LLC Fenbendazole (Standard) | 43210-67-9 | 99.9% | 100 MG
SDP
Catalog No. 5000407789
Encompass_Preferred
Fenbendazole (Standard) is an analytical standard intended for research and analytical applications. It is an orally active benzimidazole anthelmintic agent with a broad antiparasitic range, primarily acting as a microtubule destabilizing agent.
  • Binds to tubulin and disrupts microtubule equilibrium
  • Stabilizes transcriptional activator HIF-1α
  • Exhibits anti-proliferative activity, induces apoptosis, causes cell-cycle arrest, mitotic cell death, and has antitumor activity
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-07789 Supplier Medchemexpress LLC Supplier No. HYB0413R100MG
Fenbendazole (Standard) is an analytical standard intended for research and analytical applications. It is an orally active benzimidazole anthelmintic agent with a broad antiparasitic range, primarily acting as a microtubule destabilizing agent.
  • Binds to tubulin and disrupts microtubule equilibrium
  • Stabilizes transcriptional activator HIF-1α
  • Exhibits anti-proliferative activity, induces apoptosis, causes cell-cycle arrest, mitotic cell death, and has antitumor activity
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS