Learn More
Medchemexpress LLC Fenbendazole (Standard) | 43210-67-9 | 99.9% | 25 MG
SDP
Catalog No. 5000450578
Encompass_Preferred
Fenbendazole (Standard) is an analytical standard and orally active benzimidazole anthelmintic agent with a broad antiparasitic range. It functions as a microtubule destabilizing agent, binding to tubulin and disrupting microtubule equilibrium in helminthes. It also stabilizes HIF-1α, shows anti-proliferative activity, induces apoptosis, and causes cell-cycle arrest, with demonstrated antitumor activity in mice xenografted with wild-type p53.
  • Analytical standard for research and analytical applications
  • Orally active benzimidazole anthelmintic agent
  • Broad antiparasitic range
  • Microtubule destabilizing agent
  • Disrupts tubulin microtubule equilibrium
  • Stabilizes HIF-1α
  • Exhibits anti-proliferative and antitumor activity
  • Induces apoptosis and cell-cycle arrest
  • Applied in qualitative, quantitative, and methodological research (HPLC, GC, MS)
Catalog No. 50-004-50578 Supplier Medchemexpress LLC Supplier No. 25MGHYB0413R
Fenbendazole (Standard) is an analytical standard and orally active benzimidazole anthelmintic agent with a broad antiparasitic range. It functions as a microtubule destabilizing agent, binding to tubulin and disrupting microtubule equilibrium in helminthes. It also stabilizes HIF-1α, shows anti-proliferative activity, induces apoptosis, and causes cell-cycle arrest, with demonstrated antitumor activity in mice xenografted with wild-type p53.
  • Analytical standard for research and analytical applications
  • Orally active benzimidazole anthelmintic agent
  • Broad antiparasitic range
  • Microtubule destabilizing agent
  • Disrupts tubulin microtubule equilibrium
  • Stabilizes HIF-1α
  • Exhibits anti-proliferative and antitumor activity
  • Induces apoptosis and cell-cycle arrest
  • Applied in qualitative, quantitative, and methodological research (HPLC, GC, MS)