Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fenbendazole, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18602651 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-651 25 kg Amber Glass Bottle
18-602-648 25 g Amber Glass Bottle
18-602-649 100 g Amber Glass Bottle
18-602-650 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-651 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F135925KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

F1359, 43210-67-9, C15H13N3O2S

Spectrum™ Chemical Fenbendazole, USP is used as a broad spectrum benzimidazole anthelmintic that guards against gastrointestinal parasites in animals, mostly cattle, sheep, dogs, cats and rabbits. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 43210-67-9
Molecular Formula C15H13N3O2S
Molecular Weight (g/mol) 299.35
InChI Key HDDSHPAODJUKPD-UHFFFAOYSA-N
IUPAC Name methyl N-[6-(phenylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 98 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Fenbendazole, For Vet Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.