Learn More
Cayman Chemical Fenebrutinib, Purity: ≥98%, CAS: 1434048-34-6, 10 mg
SDP
Catalog No. 502985032
Encompass_Preferred
A BTK inhibitor (Kis = 0.91, 1.6, 1.3, 12.6, and 3.4 nM for wild-type BTK, BTKC481S, BTKC481R, BTKT474I, and BTKT474M, respectively); selective for BTK over a panel of 287 additional kinases at 1 µM; inhibits anti-IgM antibody-induced BTK phosphorylation in B cells (IC50 = 3.1 nM), as well as anti-IgM- or CD40L-induced proliferation of B cells (IC50s = 1.2 and 1.4 nM, respectively); reduces ankle thickness and cartilage damage in a rat model of collagen-induced arthritis
Catalog No. 50-298-5032 Supplier Cayman Chemical Supplier No. 2134610 MG
A BTK inhibitor (Kis = 0.91, 1.6, 1.3, 12.6, and 3.4 nM for wild-type BTK, BTKC481S, BTKC481R, BTKT474I, and BTKT474M, respectively); selective for BTK over a panel of 287 additional kinases at 1 µM; inhibits anti-IgM antibody-induced BTK phosphorylation in B cells (IC50 = 3.1 nM), as well as anti-IgM- or CD40L-induced proliferation of B cells (IC50s = 1.2 and 1.4 nM, respectively); reduces ankle thickness and cartilage damage in a rat model of collagen-induced arthritis