Learn More
Cayman Chemical Fenclofenac, Purity: ≥95%, CAS: 34645-84-6, 10 mg
SDP
Catalog No. 502978601
Encompass_Preferred
An NSAID and COX inhibitor (IC50s = 7 and 4 µM for COX-1 and COX-2, respectively); reduces paw volume in a rat model of adjuvant-induced polyarthritis at 3, 15, and 75 mg/kg; induces antinociception in rats (ED50 = 54.3 mg/kg); reduces carrageenan-induced paw edema in rats at 20-180 mg/kg
Catalog No. 50-297-8601 Supplier Cayman Chemical Supplier No. 4170910 MG
An NSAID and COX inhibitor (IC50s = 7 and 4 µM for COX-1 and COX-2, respectively); reduces paw volume in a rat model of adjuvant-induced polyarthritis at 3, 15, and 75 mg/kg; induces antinociception in rats (ED50 = 54.3 mg/kg); reduces carrageenan-induced paw edema in rats at 20-180 mg/kg