Learn More
Medchemexpress LLC Fenclofenac 50mg | 34645-84-6 | 297.13 | 50 MG
SDP
Catalog No. 5000229297
Encompass_Preferred
Fenclofenac (CAS 34645-84-6) is a nonsteroidal anti-inflammatory research compound provided as a high-purity reference standard for biochemical and pharmacological studies. It is supplied in small pack sizes and includes supporting documentation such as a certificate of analysis and a safety data sheet.
  • High purity 99.09%.
  • Available in small pack sizes including 50 mg.
  • Molecular formula C14H10Cl2O3; molecular weight 297.13.
  • Provided with datasheet, COA, and SDS for research use.
  • Suitable as a reference standard for pharmacology and analytical studies.
Catalog No. 50-002-29297 Supplier Medchemexpress LLC Supplier No. HY11480650MG
Fenclofenac (CAS 34645-84-6) is a nonsteroidal anti-inflammatory research compound provided as a high-purity reference standard for biochemical and pharmacological studies. It is supplied in small pack sizes and includes supporting documentation such as a certificate of analysis and a safety data sheet.
  • High purity 99.09%.
  • Available in small pack sizes including 50 mg.
  • Molecular formula C14H10Cl2O3; molecular weight 297.13.
  • Provided with datasheet, COA, and SDS for research use.
  • Suitable as a reference standard for pharmacology and analytical studies.