Learn More
Medchemexpress LLC Fenclonine (4-Chloro-DL-phenylalanine) | 7424-00-2 | 99.9% | 1 ML
SDP
Catalog No. 5000412445
Encompass_Preferred
Fenclonine is a selective and irreversible tryptophan hydroxylase inhibitor, which is a rate-limiting enzyme in the biosynthesis of serotonin. Fenclonine can be used in carcinoid syndrome research.
  • Selective and irreversible tryptophan hydroxylase inhibitor
  • Can be used in carcinoid syndrome research
  • Can be used in animal modeling to create insomnia models
Catalog No. 50-004-12445 Supplier Medchemexpress LLC Supplier No. HYB136810MM/1ML
Fenclonine is a selective and irreversible tryptophan hydroxylase inhibitor, which is a rate-limiting enzyme in the biosynthesis of serotonin. Fenclonine can be used in carcinoid syndrome research.
  • Selective and irreversible tryptophan hydroxylase inhibitor
  • Can be used in carcinoid syndrome research
  • Can be used in animal modeling to create insomnia models