Learn More
TARGETMOL CHEMICALS INC Fenclozine Maleate 1MG
SDP
Catalog No. 502879756
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fenclozine Maleate (S 1429) is a potential histamine H1 receptor antagonist , which can be used to study fever, inflammation and pain. Purity 98.47%
Catalog No. 50-287-9756 Supplier TARGETMOL CHEMICALS INC Supplier No. T839751MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Fenclozine Maleate (S 1429) is a potential histamine H1 receptor antagonist , which can be used to study fever, inflammation and pain. Purity 98.47%