Learn More
Cayman Chemical Fendiline (hydrochloride), 13636-18-5, 1 g
SDP
Supplier:  Cayman Chemical 172951
Encompass_Preferred
Molecular Formula C23H25N • HCl, Purity ≥95%, Formula Weight 351.9; An 2-adrenergic receptor antagonist (Kd = 2.6 M) and an L-type calcium channel blocker (IC50 = 17 M) well-known for its coronary vasodilator effects; inhibits K-Ras plasma membrane localization (IC50 = 9.64 M), blocking the proliferation of K-Ras-transformed tumor cells.
Catalog No. NC0986566