Learn More
Medchemexpress LLC Fendiline (hydrochloride) | 13636-18-5 | 99.8% | 1 ML
SDP
Catalog No. 5000408971
Encompass_Preferred
Fendiline hydrochloride is a diphenylalkylamine type of antianginal agent that acts as an L-type calcium channel blocker with an IC50 of 17 μM. It is also identified as a selective K-Ras inhibitor, showing no effect on H-Ras and N-Ras. It inhibits K-Ras plasma membrane localization (IC50 of 9.64 μM) and K-Ras signal output. Additionally, Fendiline hydrochloride is a STING agonist and has demonstrated the ability to inhibit the growth of multiple refractory cold tumors. This product is provided as a 10 mM solution in 1 mL of DMSO with a purity of 99.78%.
  • Acts as an L-type calcium channel blocker
  • Functions as a selective K-Ras inhibitor
  • Inhibits K-Ras plasma membrane localization
  • Suppresses K-Ras signal output
  • Prevents proliferation of various cancer cell lines
  • Serves as a STING agonist
  • Inhibits growth of refractory cold tumors
Catalog No. 50-004-08971 Supplier Medchemexpress LLC Supplier No. HYB098410MM/1ML
Fendiline hydrochloride is a diphenylalkylamine type of antianginal agent that acts as an L-type calcium channel blocker with an IC50 of 17 μM. It is also identified as a selective K-Ras inhibitor, showing no effect on H-Ras and N-Ras. It inhibits K-Ras plasma membrane localization (IC50 of 9.64 μM) and K-Ras signal output. Additionally, Fendiline hydrochloride is a STING agonist and has demonstrated the ability to inhibit the growth of multiple refractory cold tumors. This product is provided as a 10 mM solution in 1 mL of DMSO with a purity of 99.78%.
  • Acts as an L-type calcium channel blocker
  • Functions as a selective K-Ras inhibitor
  • Inhibits K-Ras plasma membrane localization
  • Suppresses K-Ras signal output
  • Prevents proliferation of various cancer cell lines
  • Serves as a STING agonist
  • Inhibits growth of refractory cold tumors