Learn More
Medchemexpress LLC Fendiline hydrochloride | 13636-18-5 | 99.8% | 500 MG
SDP
Catalog No. 5000408400
Encompass_Preferred
Fendiline hydrochloride is an antianginal agent of the diphenylalkylamine type, functioning as an L-type calcium channel blocker with an IC50 of 17 μM. It also serves as a selective K-Ras inhibitor, showing no effect on H-Ras and N-Ras. Furthermore, this compound acts as a STING agonist, capable of inhibiting the growth of multiple refractory cold tumors.
Catalog No. 50-004-08400 Supplier Medchemexpress LLC Supplier No. HYB0984500MG
Fendiline hydrochloride is an antianginal agent of the diphenylalkylamine type, functioning as an L-type calcium channel blocker with an IC50 of 17 μM. It also serves as a selective K-Ras inhibitor, showing no effect on H-Ras and N-Ras. Furthermore, this compound acts as a STING agonist, capable of inhibiting the growth of multiple refractory cold tumors.