Learn More
Selleck Chemical LLC Fendiline hydrochloride S5279-1g
SDP
Catalog No. 503112210
Encompass_Preferred
Fendiline hydrochloride is the hydrochloride salt form of Fendiline which is an L-type calcium channel blocker and also a specific inhibitor of K-Ras plasma membrane targeting with no detectable effect on the localization of H- and N-Ras
Catalog No. 50-311-2210 Supplier Selleck Chemical LLC Supplier No. S52791G
Fendiline hydrochloride is the hydrochloride salt form of Fendiline which is an L-type calcium channel blocker and also a specific inhibitor of K-Ras plasma membrane targeting with no detectable effect on the localization of H- and N-Ras