Learn More
TARGETMOL CHEMICALS INC FENEBRUTINIB 10MG
SDP
Catalog No. 502578809
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fenebrutinib (GDC-0853) is a selective and noncovalent Btk inhibitor (Ki 0.91 nM). purity: 98%
Catalog No. 50-257-8809 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ024210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fenebrutinib (GDC-0853) is a selective and noncovalent Btk inhibitor (Ki 0.91 nM). purity: 98%