Learn More
MEDILUMINE INC MediLumine Inc. Fenestra HDVC, 2 ml
Catalog No. NC3234621
Encompass_Preferred
Fenestra HDVC is an iodinated nanoemulsion formulated for microCT imaging of vasculature, liver and spleen. The product has twice the dose of iodine (100 mgI/ml) compared to previous Fenestra formulations allowing researchers to reduce the injected dose. Fenestra HDVC circulates in blood for 24 hours and the selectively targets hepatocytes for functional and anatomical imaging, staging and monitoring fatty liver disease, quantifying liver tumor burdens. Fenestra HDVC remains in liver for many months after a single injection and enabling longitudinal studies of liver diseases.
Catalog No. NC3234621 Supplier MEDILUMINE INC Supplier No. HDVC121
Fenestra HDVC is an iodinated nanoemulsion formulated for microCT imaging of vasculature, liver and spleen. The product has twice the dose of iodine (100 mgI/ml) compared to previous Fenestra formulations allowing researchers to reduce the injected dose. Fenestra HDVC circulates in blood for 24 hours and the selectively targets hepatocytes for functional and anatomical imaging, staging and monitoring fatty liver disease, quantifying liver tumor burdens. Fenestra HDVC remains in liver for many months after a single injection and enabling longitudinal studies of liver diseases.