Learn More
Roboz Surgical Fenestrated forceps, 3.5" long
SDP
Catalog No. NC1373685
Encompass
The external diameter of the round fenestrated tips is 6 mm. The internal diameter of the fenestration is 5.5 mm. The tips are not serrated.
This item is not returnable. View return policy
Catalog No. NC1373685 Supplier Roboz Surgical Supplier No. RS5162
The external diameter of the round fenestrated tips is 6 mm. The internal diameter of the fenestration is 5.5 mm. The tips are not serrated.