Learn More
TARGETMOL CHEMICALS INC FENGABINE 5MG
SDP
Catalog No. 502580629
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Fengabine (SL-79229) is a GABA receptor agonist with antidepressant activity and is used to treat depression. purity: 98%
Catalog No. 50-258-0629 Supplier TARGETMOL CHEMICALS INC Supplier No. T273105MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Fengabine (SL-79229) is a GABA receptor agonist with antidepressant activity and is used to treat depression. purity: 98%